Anti-CLEC4D Rabbit Polyclonal Antibody
PRSI59-256
New Product
- Antibody type:Primary
- Antigen name:C-type lectin domain family 4 member D
- Antigen symbol:CLEC4D
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:126 - 153 aa
- Form:Liquid
- Gene ID:338339
- Modification:None
- Molecular weight:25 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4°C
- Immunogen:This CLEC4D antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 126 - 153 amino acids from the Central region of human CLEC4D.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: CLEC4D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 126-153
Host: Rabbit
Isotype: Ig
Reactivity: Human