Order Entry
Czech Republic
ContactUsLinkComponent
Human Recombinant NGAL (from E. coli)
Human Recombinant NGAL (from E. coli)
Catalog # PRSI92-248
Supplier:  ProSci Inc.
CAS Number:  
Human Recombinant NGAL (from E. coli)
Catalog # PRSI92-248
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    0,05 mg
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimise freeze-thaw cycles.
  • Endotoxin content:
    <0,1 ng/µg (1 IEU/µg) as determined by LAL test
  • Protein synonyms:
    25 kDa alpha-2-microglobulin-related subunit of MMP-9|Neutrophil gelatinase-associated lipocalin|Lipocalin-2|HNL|LCN2|Siderocalin LCN2|NGAL|Oncogene 24p3|p25
  • Protein/peptide name:
    NGAL
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    21,8 kDa
  • Sequence:
    Gln21-Gly198
  • Formulation:
    Supplied as a 0,2 µm filtered solution of PBS, 50% glycerol, pH 7,4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 µg/ml. Dissolve the Lyophilised protein in ddH₂O. Please aliquot the reconstituted solution to minimise freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Neutrophil gelatinase-associated lipocalin(LCN2) is a secreted protein and belongs to the calycin superfamily. This protein is released from injured tubular cells after various damaging stimuli, is already known by nephrologists as one of the most promising biomarkers of incoming Acute Kidney Injury (AKI). Recent evidence also suggests its role as a biomarker in a variety of other renal and non-renal conditions. Moreover, recent studies seem to suggest a potential involvement of this factor also in the genesis and progression of chronic kidney diseases. NGAL is the first known mammalian protein which specifically binds organic molecules called siderophores, which are high-affinity iron chelators. NGAL, first known as an antibacterial factor of natural immunity, and an acute phase protein, is currently one of the most interesting and enigmatic proteins involved in the process of tumor development. acting as an intracellular iron carrier and protecting MMP9 from proteolytic degradation, NGAL has a clear pro-tumoral effect, as has already been observed in different tumors (e.g. breast, stomach, oesophagus, brain) in humans. In thyroid carcinomas, NGAL is strongly induced by NF-kB, an important factor involved both in tumor growth and in the link between chronic inflammation and neoplastic development. Thus, Lipocalin-2 (LCN2/NGAL) has been implicated in a variety of processes including cell differentiation, proliferation, survival and morphogenesis.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays.

Caution: For research use only.