Order Entry
Czech Republic
ContactUsLinkComponent
Human Recombinant Bc1-w (from E. coli)
Human Recombinant Bc1-w (from E. coli)
Catalog # PRSI91-088
Supplier:  ProSci Inc.
CAS Number:  
Human Recombinant Bc1-w (from E. coli)
Catalog # PRSI91-088
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    0,05 mg
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimise freeze-thaw cycles.
  • Endotoxin content:
    <0,1 ng/µg (1 IEU/µg) as determined by LAL test
  • Protein synonyms:
    Bcl-2-like Protein 2|Bcl2-L-2|BCL2L2|Bcl-2-Llike Protein 2|Apoptosis Regulator Bcl-W|KIAA0271|BCLW
  • Protein/peptide name:
    Bc1-w
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    19,87 kDa
  • Sequence:
    Ala2-Thr172
  • Formulation:
    Supplied as a 0,2 µm filtered solution of 25 mM HEPES, 100 mM KCl, 20% Glycerol, pH 7,5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 µg/ml. Dissolve the Lyophilised protein in ddH₂O. Please aliquot the reconstituted solution to minimise freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Bcl-2-like protein 2 (BCL2L2) belongs to the Bcl-2 family. BCL2L2 is highly expressed in thebrain, spinal cord, testis, pancreas, heart, spleen, and mammary glands. BCL2L2 is a peripheral membrane protein containing three motifs, BH1, BH2 and BH4. The BH4 motif appears to be involved in the anti-apoptotic function. The BH1 and BH2 motifs form a hydrophobic groove which acts as a docking site for the BH3 domain of some pro-apoptotic proteins. BCL2L2 promotes cell survival and blocks dexamethasone-induced apoptosis. Furthermore, BCL2L2 mediates survival of postmitotic Sertoli cells by suppressing death-promoting activity of BAX.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays.

Caution: For research use only.