Anti-HLA-DRB4 Rabbit Polyclonal Antibody
Catalog # PRSI57-203
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:HLA class II histocompatibility antigen, DR beta 4 chain
- Antigen symbol:HLA-DRB4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:43-72 aa
- Form:Liquid
- Gene ID:3126
- Modification:None
- Molecular weight:30 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,48 mg/ml
- Shipping temperature:4 °C
- Immunogen:This HLA-DRB4 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 43-72 amino acids from the N-terminal region of human HLA-DRB4
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000
Type: Primary
Antigen: HLA-DRB4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 43-72
Host: Rabbit
Isotype: Ig
Reactivity: Human