Anti-SLC43A2 Rabbit Polyclonal Antibody
Catalog # PRSI55-026
Supplier: ProSci Inc.
Specifications
- Druh protilátky:Primární
- Název antigenu:Large neutral amino acids transporter small subunit 4
- Symbol antigenu:SLC43A2
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- Průtoková cytometrie:Yes
- Hostitel:Rabbit
- Western blot:Yes
- Epitop:Amino Acid Range = 236 - 265
- Formulář:Liquid
- Identifikační číslo genu:Q8N370
- Synonyma antigenu:SLC43A2
- Modifikace:None
- Skladovací pufr:Supplied in PBS with 0,09% (W/V) sodium azide.
- Molekulární hmotnost:63 kDa
- Skladovací teplota:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Koncentrace:0,5 mg/ml
- Transportní teplota:4 °C
- Imunogen:This SLC43A2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 236 - 265 amino acids from the Central region of human SLC43A2
- Čištění:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Bal.:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000. For FACS starting dilution is: 1:10~50.
Type: Primary
Antigen: SLC43A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 236-265
Host: Rabbit
Isotype:
Reactivity: Human;Mouse