Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-FCER1A Mouse Monoclonal Antibody [clone: AER-37 (CRA1)] (FITC (Fluorescein Isothiocyanate))
Catalog # PRSI77-172
Supplier:  ProSci Inc.
Anti-FCER1A Mouse Monoclonal Antibody [clone: AER-37 (CRA1)] (FITC (Fluorescein Isothiocyanate))
Catalog # PRSI77-172
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide
  • Antigen symbol:
    FCER1A
  • Clonality:
    Monoclonal
  • Clone:
    AER-37 (CRA1)
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b kappa
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    P12319
  • Antigen synonyms:
    FCER1A
  • Storage buffer:
    Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
  • Storage temperature:
    The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
  • Shipping temperature:
    4 °C
  • Purification:
    The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
  • Pk:
    25 Tests

Specifications

About this item

The AER-37 (CRA1) antibody binds to the FC epsilon receptor I alpha subunit (FceR1a), a 50 kDa transmembrane protein. It is part of a tetrameric receptor complex composed from one alpha subunit, one beta subunit, and two gamma subunits. The FceR1a binds and is upregulated by the presence of IgE. It plays a role in prompting the allergic reactions mediated by IgE.

Type: Primary
Antigen: FCER1A
Clonality: Monoclonal
Clone: AER-37 (CRA1)
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human