Anti-Jag1 Hamster Monoclonal Antibody [clone: HMJ1-29] (PE (Phycoerythrin))
Catalog # PRSI76-995
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:jagged 1
- Antigen symbol:Jag1
- Clonality:Monoclonal
- Clone:HMJ1-29
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Hamster
- Isotype:IgG
- Reactivity:Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:Q9QXX0
- Antigen synonyms:Jag1
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:0,025 mg
Specifications
About this item
The HMJ1-29 monoclonal antibody reacts with mouse and rat CD339 (Jagged 1), a type 1 transmembrane Notch receptor ligand. It is expressed by macrophages and certain stromal, epithelial, dendritic, and thymic lymphoid cells. It has been reported to be play a role in Treg induction, Th2 polarization, neurogenesis, and cardiovascular development.
Type: Primary
Antigen: Jag1
Clonality: Monoclonal
Clone: HMJ1-29
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse