Anti-CD3E Mouse Monoclonal Antibody [clone: SK7] (APC (Allophycocyanin))
Catalog # PRSI76-304
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD3e molecule, epsilon (CD3-TCR complex)
- Antigen symbol:CD3E
- Clonality:Monoclonal
- Clone:SK7
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Gene ID:P07766
- Antigen synonyms:CD3E
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:100 Tests
Specifications
About this item
The SK7 monoclonal antibody specifically reacts with the epsilon chain of the CD3/T lymphocyte antigen receptor complex. The CD3 complex contains gamma, delta, and epsilon chains, and it is part of the TCR complex, expressed by all mature T lymphocytes and by the thymocyte lineage. CD3 enhances the antigen recognition by signal transduction. The SK7 antibody is reported to have a mitogenic effect on the majority of peripheral blood T cells in the presence of functional monocytes.
Type: Primary
Antigen: CD3E
Clonality: Monoclonal
Clone: SK7
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human