Anti-Cd8a Rat Monoclonal Antibody [clone: 53-6.7] (FITC (Fluorescein Isothiocyanate))
Catalog # PRSI76-684
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD8 antigen, alpha chain
- Antigen symbol:Cd8a
- Clonality:Monoclonal
- Clone:53-6.7
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a kappa
- Cross adsorption:No
- Form:liquid
- Gene ID:P01731
- Antigen synonyms:Cd8a
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: Cd8a
Clonality: Monoclonal
Clone: 53-6.7
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse