Anti-Cd3d Mouse Monoclonal Antibody [clone: G4.18] (FITC (Fluorescein Isothiocyanate))
Catalog # PRSI76-237
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD3 molecule, delta
- Antigen symbol:Cd3d
- Clonality:Monoclonal
- Clone:G4.18
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG3 kappa
- Cross adsorption:No
- Form:liquid
- Gene ID:P19377
- Antigen synonyms:Cd3d
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:0,05 mg
Specifications
About this item
The G4.18 monoclonal antibody specifically reacts with rat CD3, a T cell lineage marker and associated with the T cell receptor (TCR). CD3 is involved in antigen recognition, cell activation, and signal transduction and is found on subsets of NK-T, peripheral T, dendritic epidermal T, and thymocyte cells. Immobilized G4.18 antibody activates T cells in vitro and in vivo treatments of the G4.18 antibody prevents cardiac and skin allograft rejection.
Type: Primary
Antigen: Cd3d
Clonality: Monoclonal
Clone: G4.18
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG3 kappa
Reactivity: Rat