Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Kdr Rat Monoclonal Antibody [clone: Avas12a1] (PE (Phycoerythrin))
Anti-Kdr Rat Monoclonal Antibody [clone: Avas12a1] (PE (Phycoerythrin))
  PRSI76-992
New Product
Anti-Kdr Rat Monoclonal Antibody [clone: Avas12a1] (PE (Phycoerythrin))
  PRSI76-992
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    kinase insert domain protein receptor
  • Antigen symbol:
    Kdr
  • Clonality:
    Monoclonal
  • Clone:
    Avas12a1
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2a kappa
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    P35918
  • Antigen synonyms:
    Kdr
  • Storage buffer:
    Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
  • Storage temperature:
    The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
  • Shipping temperature:
    4 °C
  • Purification:
    The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
  • Pk:
    0,2 mg

 

 

The Avas12a1 monoclonal antibody specifically reacts with mouse CD309, also known as fetal liver kinase-1 (Flk-1) or the vascular endothelial growth factor receptor 2 (VEGFR2). CD309 is a receptor for VEGF and VEGFC and is expressed in endothelial cells in embryonic and adult tissue and a requirement for the development of vascular endothelial and hematopoietic cells.

Type: Primary
Antigen: Kdr
Clonality: Monoclonal
Clone: Avas12a1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse