Anti-Ncr1 Rat Monoclonal Antibody [clone: 29A1.4] (PE (Phycoerythrin))
PRSI76-982
New Product
- Antibody type:Primary
- Antigen name:natural cytotoxicity triggering receptor 1
- Antigen symbol:Ncr1
- Clonality:Monoclonal
- Clone:29A1.4
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2b kappa
- Cross adsorption:No
- Form:liquid
- Gene ID:Q8C567
- Antigen synonyms:Ncr1
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:0,025 mg
The 29A1.4 antibody specifically reacts with CD335 (NKp46), a 46 kDa natural cytotoxicity receptor family molecule. It is expressed in only NK and a rare subset of NK-like T cells, not including the CD1d-restricted NKT division. CD335 is active during NK lysis of pathogen-infected and tumor cells.The 29A1.4 in vitro activates NK cells, but does not deplete NK cells in vivo.
Type: Primary
Antigen: Ncr1
Clonality: Monoclonal
Clone: 29A1.4
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse