Anti-CLCN2 Rabbit Polyclonal Antibody
Katalogové číslo PRSI58-678
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Chloride channel protein 2
- Antigen symbol:CLCN2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:209 - 237 aa
- Form:Liquid
- Gene ID:1181
- Modification:None
- Molecular weight:99 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,45 mg/ml
- Shipping temperature:4 °C
- Immunogen:This CLCN2 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 209-237 amino acids from the N-terminal region of human CLCN2
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: CLCN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 209-237
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse