Anti-NKAIN2 Rabbit Polyclonal Antibody
Catalog # PRSI58-633
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Sodium/potassium-transporting ATPase subunit beta-1-interacting protein 2
- Antigen symbol:NKAIN2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:106 - 134 aa
- Form:Liquid
- Gene ID:154215
- Modification:None
- Molecular weight:24 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This NKAIN2 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 106-134 amino acids from the Central region of human NKAIN2
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: NKAIN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 106-134
Host: Rabbit
Isotype: Ig
Reactivity: Human