Anti-VCAM1 Rabbit Polyclonal Antibody
Catalog # PRSI57-990
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Vascular cell adhesion protein 1
- Antigen symbol:VCAM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:295 - 322 aa
- Form:Liquid
- Gene ID:7412
- Modification:None
- Molecular weight:81 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Shipping temperature:4 °C
- Immunogen:This CD106 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 295-322 amino acids from the Central region of human CD106
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: VCAM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 295-322
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse