Anti-ZCCHC4 Rabbit Polyclonal Antibody
Catalog # PRSI13-458
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:zinc finger CCHC-type containing 4
- Antigen symbol:ZCCHC4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:29063
- Antigen synonyms:GRF-type containing 4|ZGRF4|DHHC domain-containing zinc finger protein|HSPC052|zinc finger|rRNA N6-adenosine-methyltransferase ZCCHC4|CCHC domain containing 4|zinc finger CCHC domain-containing protein 4
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 50-220 of human ZCCHC4 (NP_079212.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: ZCCHC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse