Anti-RAN Rabbit Polyclonal Antibody
Catalog # PRSI13-347
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:RAN, member RAS oncogene family
- Antigen symbol:RAN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:5901
- Antigen synonyms:RanGTPase|TC4|OK/SW-cl.81|Gsp1|OTTHUMP00000166078|GTP-binding nuclear protein Ran|RAN|member RAS oncogene|guanosine triphosphatase Ran|ARA24|GTPase Ran|androgen receptor-associated protein 24|member RAS oncogene family
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-216 of human RAN (NP_006316.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: RAN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey