Anti-Spike Protein Rabbit Recombinant Antibody [clone: 8F8C0]
Catalog # PRSI11-093
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Surface glycoprotein
- Antigen symbol:Spike Protein
- Clonality:Recombinant
- Clone:8F8C0
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Virus
- Form:Liquid
- Gene ID:43740568
- Antigen synonyms:Spike glycoprotein
- Modification:None
- Storage buffer:Supplied as a 0,2 µm filtered solution in PBS, pH 7,4
- Concentration:1 mg/ml
- Shipping temperature:20
- Immunogen:HEK293 derived SARS-COV-2 S1+S2 ECD (S-ECD)
- Purification:Affinity purified, endotoxin level: <1,0 EU/µg of the protein by LAL method
- Pack type:Vial
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: Spike protein
Clonality: Recombinant
Clone: 8F8C0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Virus