Order Entry
Czech Republic
ContactUsLinkComponent
Anti-LY-2951742 Recombinant Antibody
Anti-LY-2951742 Recombinant Antibody
Catalog # PRSI10-772
Supplier:  ProSci Inc.
Anti-LY-2951742 Recombinant Antibody
Catalog # PRSI10-772
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Recombinant
  • Conjugation:
    Unconjugated
  • Isotype:
    IgG1 kappa
  • Antigen synonyms:
    MOXR-0916|RG-7888|MOXR0916|RO7021608
  • Modification:
    None
  • Storage buffer:
    PBS buffer pH 7,5
  • Storage temperature:
    Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2…8 °C for 1 - 2 weeks. Store at –20 °C for 12 months. Store at –80 °C for long term storage.
  • Concentration:
    Batch dependent
  • Shipping temperature:
    20
  • Immunogen:
    Humanised/TNFRSF4/CD134 [Homo sapiens]
  • Purification:
    >95%
  • Pack type:
    Vial
  • Pk:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1-kappa
Reactivity: