Anti-PDE6C Rabbit Polyclonal Antibody
Catalog # PRSI64-082
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Cone cGMP-specific 3',5'-cyclic phosphodiesterase subunit alpha'
- Antigen symbol:PDE6C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Epitope:271-300 aa
- Form:Liquid
- Gene ID:5146
- Modification:None
- Molecular weight:99 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This PDE6C antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 271-300 amino acids from the Central region of human PDE6C.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
Specifications
About this item
For IHC-P starting dilution is: 1:50~100.
Type: Primary
Antigen: PDE6C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 271-300
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse