Anti-HSPB6 Rabbit Polyclonal Antibody
Catalog # PRSI64-037
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Heat shock protein beta-6
- Antigen symbol:HSPB6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:91-120 aa
- Form:Liquid
- Gene ID:126393
- Modification:None
- Molecular weight:17 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This HSPB6 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 91-120 amino acids from the Central region of human HSPB6.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: HSPB6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 91-120
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse