Anti-RPLP2 Rabbit Polyclonal Antibody
PRSI63-899
New Product
- Antibody type:Primary
- Antigen name:60S acidic ribosomal protein P2
- Antigen symbol:RPLP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:13-41 aa
- Form:Liquid
- Gene ID:6181
- Modification:None
- Molecular weight:12 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,39 mg/ml
- Shipping temperature:4 °C
- Immunogen:This RPLP2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 13-41 amino acids from the N-terminal region of human RPLP2.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: RPLP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 13-41
Host: Rabbit
Isotype: Ig
Reactivity: Human