Anti-ACY3 Rabbit Polyclonal Antibody
Katalogové číslo PRSI63-608
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:N-acyl-aromatic-L-amino acid amidohydrolase (carboxylate-forming)
- Antigen symbol:ACY3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:66-95 aa
- Form:Liquid
- Gene ID:91703
- Modification:None
- Molecular weight:35 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Shipping temperature:4 °C
- Immunogen:This ACY3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 66-95 amino acids from the Central region of human ACY3.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000. For FACS starting dilution is: 1:10~50.
Type: Primary
Antigen: ACY3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 66-95
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse