Anti-C1QBP Rabbit Polyclonal Antibody
Catalog # PRSI63-231
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Complement component 1 Q subcomponent-binding protein, mitochondrial
- Antigen symbol:C1QBP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:221-249 aa
- Form:Liquid
- Gene ID:708
- Modification:None
- Molecular weight:31 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Shipping temperature:4 °C
- Immunogen:This GC1qR (C1QBP) antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 221-249 amino acids from the C-terminal region of human GC1qR (C1QBP).
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: C1QBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 221-249
Host: Rabbit
Isotype: Ig
Reactivity: Human