Anti-SLC6A8 Rabbit Polyclonal Antibody
Catalog # PRSI61-302
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Sodium- and chloride-dependent creatine transporter 1
- Antigen symbol:SLC6A8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Form:Liquid
- Gene ID:6535
- Modification:None
- Molecular weight:71 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This SLC6A8 antibody is generated from a rabbit immunised with a KLH conjugated synthetic peptide between 180 - 212 amino acids from human SLC6A8.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:200 µl
Specifications
About this item
For WB starting dilution is: 1:2000.
Type: Primary
Antigen: SLC6A8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Ig
Reactivity: Mouse