Anti-GIGYF2 Rabbit Polyclonal Antibody
PRSI61-231
- Antibody type:Primary
- Antigen name:PERQ amino acid-rich with GYF domain-containing protein 2
- Antigen symbol:GIGYF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:26058
- Modification:None
- Molecular weight:150 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This GIGYF2 antibody is generated from a rabbit immunised with a KLH conjugated synthetic peptide between 835 - 869 amino acids from the Central region of human GIGYF2.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:200 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: GIGYF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Ig
Reactivity: Human