Anti-MT-CO3 Rabbit Polyclonal Antibody
PRSI60-323
- Antibody type:Primary
- Antigen name:Cytochrome c oxidase subunit 3
- Antigen symbol:MT-CO3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:48 - 77 aa
- Form:Liquid
- Gene ID:4514
- Modification:None
- Molecular weight:30 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This MT - CO3 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 48 - 77 amino acids from the N-terminal region of human MT - CO3.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: MT-CO3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 48-77
Host: Rabbit
Isotype: Ig
Reactivity: Human