Anti-CRHR1 Rabbit Polyclonal Antibody
PRSI23-564
- Antibody type:Primary
- Antigen name:corticotropin releasing hormone receptor 1
- Antigen symbol:CRHR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:1394
- Antigen synonyms:CRF1|corticotropin releasing hormone receptor variant 1h|CRFR|seven transmembrane helix receptor|CRF-R|CRHR|CRHR1|CRFR1|CRH-R1h|corticotropin releasing hormone receptor 1|CRHR1f
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 24-120 of human CRHR1 (NP_001138618.1)
- Purification:Affinity purification.
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: CRHR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat