Anti-MSLN Recombinant Antibody [Clone: 11-25]
Catalog # PRSI31-540
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:mesothelin
- Antigen symbol:MSLN
- Clonality:Recombinant
- Clone:11-25
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Cross adsorption:No
- Form:Liquid
- Gene ID:10232
- Antigen synonyms:Pre-pro-megakaryocyte-potentiating factor|CAK1 antigen|MSLN
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:The original antibody was generated by intraperitoneally immunizing BALB/c mice with the purified recombinant mesothelin protein
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: mesothelin
Clonality: Recombinant
Clone: 11-25
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2b kappa
Reactivity: Human