Anti-PRDM9 Recombinant Antibody [Clone: RAB-C370]
PRSI24-850
- Antibody type:Primary
- Antigen name:PR/SET domain 9
- Antigen symbol:PRDM9
- Clonality:Recombinant
- Clone:RAB-C370
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Cross adsorption:No
- Form:Liquid
- Gene ID:56979
- Antigen synonyms:PR domain zinc finger protein 9|PR domain-containing protein 9|PRDM9-A003-1459|[histone H4]-N-methyl-L-lysine20 N-methyltransferase PRDM9|EC:2.1.1.-|[histone H3]-lysine36 N-trimethyltransferase PRDM9|[histone H4]-lysine20 N-methyltransferase PRDM9|[histone H3]-lysine4 N-trimethyltransferase PRDM9|Histone-lysine N-methyltransferase PRDM9|Protein-lysine N-methyltransferase PRDM9|[histone H3]-lysine9 N-trimethyltransferase PRDM9|anti-PRDM9-RAB-C370
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was obtained by recombinant antibody (rAb) phage display recognizing PRDM9 protein under non-denaturing conditions
- Pack type:Vial
- Pk:0,2 mg
Type: Primary
Antigen: PR/SET domain 9
Clonality: Recombinant
Clone: RAB-C370
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human