Anti-CD53 Recombinant Antibody [Clone: 161-2 (53/2)]
Catalog # PRSI24-758
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:CD53 molecule
- Antigen symbol:CD53
- Clonality:Recombinant
- Clone:161-2 (53/2)
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Cross adsorption:No
- Form:Liquid
- Gene ID:963
- Antigen synonyms:Leukocyte surface antigen CD53|Tetraspanin-25|Tspan-25|Cell surface glycoprotein CD53
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was raised by immunizng mice with stimulated human leukocytes
- Tested applications:Functional assays
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: CD53 molecule
Clonality: Recombinant
Clone: 161-2 (53/2)
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human