Anti-Vsir Recombinant Antibody [Clone: 13F3]
Catalog # PRSI24-714
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:V-set immunoregulatory receptor
- Antigen symbol:Vsir
- Clonality:Recombinant
- Clone:13F3
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Isotype:IgG lambda
- Cross adsorption:No
- Form:Liquid
- Gene ID:74048
- Antigen synonyms:Platelet receptor Gi24|V-domain Ig suppressor of T cell activation|Vsir|V-set immunoregulatory receptor|V-type immunoglobulin domain-containing suppressor of T-cell activation|PD-1H|Dies1|V-set domain-containing immunoregulatory receptor
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was raised by immunising Armenian hamsters with EL4 cells overexpressing VISTA-RFP, and VISTA-Ig fusion protein
- Tested applications:Blocking
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: V-set immunoregulatory receptor
Clonality: Recombinant
Clone: 13F3
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG lambda
Reactivity: Mouse