Anti-Il2rb Recombinant Antibody [Clone: 5H4]
Catalog # PRSI24-694
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:interleukin 2 receptor, beta chain
- Antigen symbol:Il2rb
- Clonality:Recombinant
- Clone:5H4
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Cross adsorption:No
- Form:Liquid
- Gene ID:16185
- Antigen synonyms:High affinity IL-2 receptor subunit beta|CD122|Interleukin-2 receptor subunit beta|IL-2Rβ|p70-75|IL-2 receptor subunit beta
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was raised by immunising Lewis rats with transfected rat myeloma cell line YB2/0, expressing truncated murine IL-2R beta
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: interleukin 2 receptor, beta chain
Clonality: Recombinant
Clone: 5H4
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2a kappa
Reactivity: Mouse