Anti-Sell Recombinant Antibody [Clone: MEL-14]
Catalog # PRSI24-438
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:selectin, lymphocyte
- Antigen symbol:Sell
- Clonality:Recombinant
- Clone:MEL-14
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Cross adsorption:No
- Form:Liquid
- Gene ID:20343
- Antigen synonyms:Leu-8|Ly-22|LECAM-1|MEL14|L-selectin
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:C3H/eb mouse B lymphoma 38C-13
- Tested applications:Blocking
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: selectin, lymphocyte
Clonality: Recombinant
Clone: MEL-14
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2a kappa
Reactivity: Mouse