Anti-GCN4 Recombinant Antibody [Clone: C11L34]
PRSI24-378
- Antibody type:Primary
- Antigen name:amino acid starvation-responsive transcription factor GCN4
- Antigen symbol:GCN4
- Clonality:Recombinant
- Clone:C11L34
- Conjugation:Unconjugated
- ImmunoPrecipitation:Yes
- Isotype:IgG1 lambda
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:856709
- Antigen synonyms:General Control Protein 4
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:Biotinylated GCN4-peptide (HLENEVARLKK)
- Tested applications:Surface plasma resonance
- Pack type:Vial
- Pk:0,2 mg
Type: Primary
Antigen: amino acid starvation-responsive transcription factor GCN4
Clonality: Recombinant
Clone: C11L34
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1 lambda
Reactivity: S. cerevisiae