Order Entry
Czech Republic
ContactUsLinkComponent
Anti-BCL5 Recombinant Antibody [Clone: SAIC-23E-30]
Anti-BCL5 Recombinant Antibody [Clone: SAIC-23E-30]
Catalog # PRSI24-318
Supplier:  ProSci Inc.
Anti-BCL5 Recombinant Antibody [Clone: SAIC-23E-30]
Catalog # PRSI24-318
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    B cell CLL/lymphoma 5
  • Antigen symbol:
    BCL5
  • Clonality:
    Recombinant
  • Clone:
    SAIC-23E-30
  • Conjugation:
    Unconjugated
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    603
  • Antigen synonyms:
    Stress-responsive protein 27|Heat shock 27 kDa protein|SRP27|HSP27|HSP28|Estrogen-regulated 24 kDa protein|HSP 27|Heat shock protein beta-1|28 kDa heat shock protein
  • Modification:
    Unmodified
  • Storage buffer:
    PBS with 0,02% Proclin 300
  • Storage temperature:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    Peptide "LFDQAFGLPR" derived from Heat shock protein beta-1, involved in stress resistance and actin organisation, conjugated to KLH
  • Tested applications:
    ImmunoMRM
  • Pack type:
    Vial
  • Pk:
    0,2 mg

Specifications

About this item

Type: Primary
Antigen: B cell CLL/lymphoma 5
Clonality: Recombinant
Clone: SAIC-23E-30
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human