Anti-MOPC21 Recombinant Antibody [Clone: MOPC-21]
PRSI24-226
- Antibody type:Primary
- Clonality:Recombinant
- Clone:MOPC-21
- Conjugation:Unconjugated
- Isotype:IgG3 kappa
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:MOPC21
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Tested applications:Negative control
- Pack type:Vial
- Pk:0,2 mg
Type: Primary
Antigen: MOPC21
Clonality: Recombinant
Clone: MOPC-21
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG3 kappa
Reactivity: