Anti-CD3E Recombinant Antibody [Clone: YTH 12.5]
Catalog # PRSI24-204
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:CD3 epsilon subunit of T-cell receptor complex
- Antigen symbol:CD3E
- Clonality:Recombinant
- Clone:YTH 12.5
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG lambda
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:916
- Antigen synonyms:CD3E|CD3e|T3E|T-cell surface glycoprotein CD3 epsilon chain|T-cell surface antigen T3/Leu-4 epsilon chain|YTH12.5
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:Human CD3
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: CD3 epsilon subunit of T-cell receptor complex
Clonality: Recombinant
Clone: YTH 12.5
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG lambda
Reactivity: Human