Anti-Fluorescein (FITC) Recombinant Antibody [Clone: 4-4-20 (enhanced)]
Catalog # PRSI24-077
Dodavatel: ProSci Inc.
Specifikace
- Antibody type:Primary
- Clonality:Recombinant
- Clone:4-4-20 (enhanced)
- Conjugation:Unconjugated
- Isotype:IgG2a kappa
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:FITC|FL|fluorescyl|fluoresceine
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:Fluorescein I-bovine serum albumin (FlI-BSA)
- Tested applications:Negative control
- Pack type:Vial
- Pk:0,2 mg
Specifikace
O tomto produktu
Type: Primary
Antigen: Fluorescein (FITC)
Clonality: Recombinant
Clone: 4-4-20 (enhanced)
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2a kappa
Reactivity: