Order Entry
Czech Republic
ContactUsLinkComponent
Anti-L3MBTL1 Recombinant Antibody [Clone: RAB-C213]
Anti-L3MBTL1 Recombinant Antibody [Clone: RAB-C213]
Catalog # PRSI24-845
Supplier:  ProSci Inc.
Anti-L3MBTL1 Recombinant Antibody [Clone: RAB-C213]
Catalog # PRSI24-845
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    L3MBTL histone methyl-lysine binding protein 1
  • Antigen symbol:
    L3MBTL1
  • Clonality:
    Recombinant
  • Clone:
    RAB-C213
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    Fab fragment kappa
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    26013
  • Antigen synonyms:
    H-l(3)mbt|anti-L3MBTL1-RAB-C213|L(3)mbt protein homolog|L(3)mbt-like|L3MBTL1-A003-1453|L3MBTL1|Lethal(3)malignant brain tumor-like protein 1|Lethal 3 malignant brain tumor-like protein 1|H-l(3)mbt protein
  • Modification:
    Unmodified
  • Storage buffer:
    PBS with 0,02% Proclin 300
  • Storage temperature:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    This antibody was obtained by recombinant antibody (rAb) phage display recognizing L3MBTL1 protein under non-denaturing conditions
  • Pack type:
    Vial
  • Pk:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen: L3MBTL histone methyl-lysine binding protein 1
Clonality: Recombinant
Clone: RAB-C213
Conjugation: Unconjugated
Host: Recombinant
Isotype: Fab fragment kappa
Reactivity: Human