Order Entry
Czech Republic
ContactUsLinkComponent
Anti-KRT7 Recombinant Antibody [Clone: OV-TL 12/30]
Anti-KRT7 Recombinant Antibody [Clone: OV-TL 12/30]
Catalog # PRSI24-801
Supplier:  ProSci Inc.
Anti-KRT7 Recombinant Antibody [Clone: OV-TL 12/30]
Catalog # PRSI24-801
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    keratin 7
  • Antigen symbol:
    KRT7
  • Clonality:
    Recombinant
  • Clone:
    OV-TL 12/30
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    3855
  • Antigen synonyms:
    CK7|Keratin|Sarcolectin|type II cytoskeletal 7|Cytokeratin-7|CK-7|Type-II keratin Kb7|K7|CK 7|Keratin-7
  • Modification:
    Unmodified
  • Storage buffer:
    PBS with 0,02% Proclin 300
  • Storage temperature:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    This antibody was originally raised by immunizing BALB/c mice with ovarian carcinoma cell line OTN11
  • Pack type:
    Vial
  • Pk:
    0,2 mg

Specifications

About this item

Type: Primary
Antigen: keratin 7
Clonality: Recombinant
Clone: OV-TL 12/30
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG1 kappa
Reactivity: Human