Anti-C5AR1 Recombinant Antibody [Clone: 32-G1]
PRSI24-779
- Antibody type:Primary
- Antigen name:complement C5a receptor 1
- Antigen symbol:C5AR1
- Clonality:Recombinant
- Clone:32-G1
- Conjugation:Unconjugated
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:728
- Antigen synonyms:C5a anaphylatoxin chemotactic receptor|C5a-R|32G-1|C5a anaphylatoxin chemotactic receptor 1|C5aR|CD88|pC5aR
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was raised by immunizing mice a synthetic C5aR C-terminal phosphopeptide that contained phosphoserine instead of serine at position 334 and also at the neighboring phosphorylation sites Ser-332 and Ser-338
- Pack type:Vial
- Pk:0,2 mg
Type: Primary
Antigen: complement C5a receptor 1
Clonality: Recombinant
Clone: 32-G1
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG1 kappa
Reactivity: Human