Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Cd79b Recombinant Antibody [Clone: HM79-16]
Anti-Cd79b Recombinant Antibody [Clone: HM79-16]
Catalog # PRSI24-773
Supplier:  ProSci Inc.
Anti-Cd79b Recombinant Antibody [Clone: HM79-16]
Catalog # PRSI24-773
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD79B antigen
  • Antigen symbol:
    Cd79b
  • Clonality:
    Recombinant
  • Clone:
    HM79-16
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG lambda
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    15985
  • Antigen synonyms:
    CD79b molecule|AGM6|IGB|B29|Ig-beta
  • Modification:
    Unmodified
  • Storage buffer:
    PBS with 0,02% Proclin 300
  • Storage temperature:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    HM79-16 was initially generated by purifying CD79b from WEHI231 B cells and used for immunisation of footpads of an Armenian hamster. Hybridoma supernatants were screened for positive reactivity in surface immunofluorescence of WEHI231 and then the specificity to CD79b was tested by immuno-precipitationRecombinant anti-CD79 antibodies were engineered by cloning and sequencing the full-length IG heavy and light chains of an Armenian hamster (Cricetulus migratorius)
  • Pack type:
    Vial
  • Pk:
    0,2 mg

Specifications

About this item

Type: Primary
Antigen: CD79B antigen
Clonality: Recombinant
Clone: HM79-16
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG lambda
Reactivity: Mouse