Anti-Timd2 Recombinant Antibody [Clone: RMT2-14]
Catalog # PRSI24-629
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:T cell immunoglobulin and mucin domain containing 2
- Antigen symbol:Timd2
- Clonality:Recombinant
- Clone:RMT2-14
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- Isotype:IgG lambda
- Reactivity:Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:171284
- Antigen synonyms:TIMD2|T Cell immunoglobulin and mucin domain protein 2|T Cell Immunoglobulin Mucin-2|TIM3
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was raised by immunising Sprague Dawley rats with TIM-2-Ig consisting of the extracellular domain (aa 1-230 of mouse TIM-2) and the Fc portion of mouse IgG2a, emulsified in complete Freund's adjuvant and then fusing lymph node cells from the immunized rats with P3U1 myeloma cells
- Tested applications:Blocking,Functional Assays
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: T cell immunoglobulin and mucin domain containing 2
Clonality: Recombinant
Clone: RMT2-14
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG lambda
Reactivity: Mouse