Anti-CD40L Recombinant Antibody [Clone: AT161-10]
PRSI24-535
- Antibody type:Primary
- Clonality:Recombinant
- Clone:AT161-10
- Conjugation:Unconjugated
- ELISA:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:CD-40L|CD-40 Ligand|CD154|CD-40-L|CD40-L|CD40 ligand
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was raised by immunising mice with Chinese hamster ovary (CHO) cells expressing human CD40
- Pack type:Vial
- Pk:0,2 mg
Type: Primary
Antigen: CD40L
Clonality: Recombinant
Clone: AT161-10
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG kappa
Reactivity: Human