Anti-PD-1 Recombinant Antibody [Clone: RMP1-14]
PRSI24-531
- Antibody type:Primary
- Clonality:Recombinant
- Clone:RMP1-14
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:RMP1/14|Programmed Death 1|RMPI-14|Programmed Death Receptor 1|CD279|PDCD1|PD1
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:The original rat hybridoma was generated by immunizing Sprague Dawley rats with mouse PD-1-transfectked BHK cells A P3U1 myeloma was used as the fusion partne
- Tested applications:Blocking
- Pack type:Vial
- Pk:0,2 mg
Type: Primary
Antigen: PD-1
Clonality: Recombinant
Clone: RMP1-14
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG2a kappa
Reactivity: Mouse