Anti-C3 Recombinant Antibody [Clone: S3 12D]
Catalog # PRSI24-395
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:complement C3
- Antigen symbol:C3
- Clonality:Recombinant
- Clone:S3 12D
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG kappa
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:718
- Antigen synonyms:PIG61|HER1|mENA|ERBB|NISBD2|Epidermal growth factor receptor|ERBB1
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:Human EGF
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: complement C3
Clonality: Recombinant
Clone: S3 12D
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG kappa
Reactivity: Human