Anti-TAG-72 Recombinant Antibody [Clone: B72.3 (Satumomab)]
Katalogové číslo PRSI24-277
Dodavatel: ProSci Inc.
Specifikace
- Antibody type:Primary
- Clonality:Recombinant
- Clone:B72.3 (Satumomab)
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:TAG-72
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:Tumour-associated glycoprotein 72 is a glycoprotein found on the surface of many cancer cells, including ovary, breast, colon, and pancreatic cell
- Tested applications:Imaging
- Pack type:Vial
- Pk:0,2 mg
Specifikace
O tomto produktu
Type: Primary,Biosimilars
Antigen: TAG-72
Clonality: Recombinant
Clone: B72.3 (Satumomab)
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG kappa
Reactivity: Human