ContactUsLinkComponent
Plastic Box with White Lid
Plastic Box with White Lid
Catalog # MULC192-3000
Supplier:  MULTIROIR
undefined
Plastic Box with White Lid
Catalog # MULC192-3000
Supplier:  MULTIROIR
Specifications
  • Type:
    Safety lock
  • Capacity:
    3 L
  • Material:
    PP
  • W×D×H:
    193×193×112 mm
  • Pk:
    72
Specifications
About this item
These combi box containers are versatile, providing ideal storage solutions for a diverse range of laboratory applications.
  • Polypropylene material
  • Watertight lid
  • Tamper-proof lid
  • Stackable box
  • Food contact quality
Ideal for laboratory environments, this versatile container ensures secure storage and transport of samples and reagents. The airtight, tamper-evident lid provides enhanced safety, while the stackable design maximises space efficiency. The opaque white polypropylene construction ensures durability and easy identification of contents.