Protein extraction reagent, BugBuster® 10X, Millipore®
70921-4
:
- Pk:50 mL
- Description:Protein extraction reagent, BugBuster® 10X
BugBuster® 10X is a concentrated formulation of the proprietary detergents employed in BugBuster® without the addition of salts or buffer components. Concentrated BugBuster® provides a flexible alternative to the ready-to-use standard 1X BugBuster®, allowing user-defined dilution and addition of buffer components. BugBuster® 10X has all of the bioprocessing benefits of standard BugBuster® plus the freedom to control pH, reagent concentration, and buffer additives necessary for maximum extraction and activity of your target protein.
Storage: 15 to 30 °C